Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ Protector™ Fiberglass Filtered Glove Boxes
Features inlet and outlet HEPA filters, 99.99% efficient on particulates
Supplier: Labconco™ 5065035
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Requires : Supporting base
Description
- One-piece molded fiberglass liner
- Clear acrylic inner and outer transfer chamber doors
- Two glove ports
- 0.3μm particulate containment <20 ng/m3
- Built-in blower with speed control capable of airflow from 15 CFM to 85 CFM
- 0.05 ppm average tracer gas levels (surpasses ASHRAE 110 standards)
- Dry powder-coated, 18 and 20 gauge steel exterior panel
- Bag-In/Bag-Out filter disposal system
- Space-saving inner and outer transfer chamber doors pivoting upward equipped with quick-latches
- Two 8 in. ID, epoxy-coated aluminum, double-grooved glove ports mounted 17 in. apart with neoprene gaskets
- Two 25-watt fluorescent lights that exceed 120 footcandles
- LCD display for real time monitoring of volume dilution rates in cubic feet per minute, air changes/minute, and main chamber pressure (inches of water)
- Interior left-hand side wall-mounted pass-through plug
- Interior right-hand side wall-mounted electrical receptacle
- With pair of neoprene gloves, 0.015 in. thick, 30 in. long
- Large viewing area, 36W x 27.7 in. H with 0.375 in. thick, laminated safety glass
- Dimensions: 63.9W x 31.0D x 45.7 in. H (162.3 x 78.7 x 116.1cm); Interior working width: 35.5 in.
Work with hazardous materials
Certifications
Exceeds ISO Class 4 conditions and achieves ISO Class 3 conditions, CE marked (International models)
Compliance
ETL listing, UL 61010-1 and CAN/CSA C22.2 No. 1010.1 conformity (North America models)
Specifications
Glove Box | |
Fluorescent (2 x 25 W) | |
10 A | |
31 in. | |
LCD | |
116.1 cm | |
Schuko Plug | |
208/230 V | |
227 kg | |
35 in. | |
88.9 cm |
HEPA | |
Fiberglass | |
CE | |
78.7 cm | |
45.7 in. | |
50/60 Hz | |
2 | |
500 lb. | |
63.9 in. | |
162.3 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction