Labconco™ Protector™ Standard Storage Cabinet

The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.

Manufacturer: Labconco™ 9900400

 View more versions of this product

Catalog No. 16-080-316

  Check Availability
Add to Cart

    Specifications

    Height (English) Base 35.5 to 36.75 in.
    Height (Metric) Base 90.17 to 93.34cm
    Load Capacity (English) Centralized 800 lb.
    Material Epoxy-Coated Steel
    Width (English) 18 in.
    Depth (English) 22 in.
    Width (Metric) 45.72cm
    Depth (Metric) 55.88cm
    Door Type Single, Right Hinge, Manual Close
    For Use With (Application) Ideal for storage and venting of small laboratory instrument and mild acid and other reagent
    Includes Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in.
    Load Capacity (Metric) Centralized 362.8kg
    Product Type Standard Storage Cabinet
    Weight (English) 90 lb.
    Weight (Metric) 40.82kg

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit