Labconco™ Protector™ Standard Storage Cabinet

The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.

Manufacturer: Labconco™ 9904400

 View more versions of this product

Catalog No. 16-080-324

  Check Availability
Add to Cart

    Specifications

    Certifications ADA
    Width (Metric) 45.72cm
    Depth (Metric) 55.88cm
    Door Type Single, Left Hinge, Manual Close
    For Use With (Application) Ideal for storage and venting of small laboratory instrument and mild acid and other reagent
    Height (English) Base 31.5 to 32.75 in.
    Height (Metric) Base 80 to 83.18cm
    Includes Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in.
    Load Capacity (English) Centralized 800 lb.
    Load Capacity (Metric) Centralized 362.8kg
    Material Epoxy-Coated Steel
    Product Type Standard Storage Cabinet
    Weight (English) 85 lb.
    Weight (Metric) 38.55kg
    Width (English) 18 in.
    Depth (English) 22 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit