Learn More
Labconco™ PuriCare™ Flow Station Replacement Filters
Replacement filters for use with PuriCare Animal Research Stations.
Manufacturer:  Labconco™ 3796201
 View more versions of this product
Catalog No. 09-039-127
Add to cart
Description
Description
Replacement filters for use with PuriCare Animal Research Stations.
Replacement Activated Carbon with Alumina Exhaust Filter is a granular activated carbon and impregnated alumina media adsorb odors, pollutants and light gases. For use with 3' PuriCare Bedding Disposal Stations.
Specifications
Specifications
PuriCare replacement filter
3 ft. PuriCare™ bedding disposal station and 6 ft. procedure station
1 lb.
Charcoal filter
Adsorb odors, pollutants and light gases
0.5kg