Learn More
Labconco™ Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet
Labconco Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet features ergonomic Inclination™ Technology and MyLogic™ Operating System.
Manufacturer:  Labconco™ 304410010
 View more versions of this product
Catalog No. 30-441-0010
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Description
Description
The Purifier Axiom Class II, Type C1 is first biological safety cabinet that can be used in recirculating Type A-mode for standard microbiological work or can be connected to exhaust system to function in Type-B mode for handling hazardous chemical vapors or radionuclides.
  • Safe and easy to operate than traditional biosafety cabinets due to efficient, well-defined airflow pattern over its work surface
  • Chem-Zone™(patent pending) increases chemical safety by creating large, clearly defined area within the enclosure
  • Provides protection for personnel, product and laboratory environment by filtering hazardous particulates and aerosols, such as agents that require Biosafety Level 1, 2 or 3 containment, w/option of venting away hazardous fumes
C1 designation is pending NSF approval
Specifications
Specifications
25.4cm
81.2cm
Antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
N/A
North America
10 in.
288.5kg
137.6cm
69 in.
32 in.
230V, 50/60Hz, 6A, Domestic
163cm
Fluorescent
Safety Cabinet
636 lb.
54.2 in.
NSF International-Listed, ETL, Modified ASHRAE 110