Labconco™ Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet

Labconco Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet features ergonomic Inclination™ Technology and MyLogic™ Operating System.

Manufacturer: Labconco™ 304411070

 View more versions of this product

Catalog No. 30-441-1070

  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product.

Please contact customer service for assistance: 1-800-766-7000.

Add to Cart

    Specifications

    Electrical Requirements 230V, 50/60Hz, 6A, International
    For Use With (Application) Applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
    Product Type Safety Cabinet
    Height (Metric) 163cm
    Certifications/Compliance NSF International-Listed, ETL, Modified ASHRAE 110,CE
    Weight (English) 636 lb.
    Weight (Metric) 288.5kg
    Sash Opening (Metric) 25.4cm,25.4cm
    Amperage 6A
    Depth (English) 32 in.
    Hertz 50/60Hz
    Includes Service fixture and Vacu-Pass Portal
    Lighting Fluorescent, UV
    Plug Type India
    Sash Opening (English) 10 in.
    Width (English) 54.2 in.
    Height (English) 69 in.
    Depth (Metric) 81.2cm
    Width (Metric) 137.6cm
    Voltage 230V
    Type Purifier Axiom C1 Biological Safety Cabinet

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit