Learn More
Labconco™ Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet
Labconco Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet features ergonomic Inclination™ Technology and MyLogic™ Operating System.
Manufacturer:  Labconco™ 304411130
 View more versions of this product
Catalog No. 30-441-1130
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Specifications Description & Specifications
Specifications
32 in.
230V, 50/60Hz, 6A, International
233cm
Fluorescent, UV
Safety Cabinet
721 lb.
54.2 in.
NSF International-Listed, ETL, Modified ASHRAE 110
25.4cm
81.2cm
Applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
Service fixture, Vacu-Pass Portal, Base Stand
Schuko
10 in.
327kg
137.6cm
96.5 in.