Labconco™ Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet

Labconco Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet features ergonomic Inclination™ Technology and MyLogic™ Operating System.

Manufacturer: Labconco™ 304410120

 View more versions of this product

Catalog No. 30-441-0120

  Check Availability
Add to Cart

    Specifications

    Electrical Requirements 100V, 50/60Hz, 12A, International
    For Use With (Application) Applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
    Plug Type N/A
    Product Type Safety Cabinet
    Certifications/Compliance NSF International-Listed, ETL, Modified ASHRAE 110
    Height (Metric) 233cm
    Depth (English) 32 in.
    Includes Base stand
    Lighting Fluorescent
    Sash Opening (English) 10 in.
    Weight (Metric) 327kg
    Width (English) 54.2 in.
    Width (Metric) 137.6cm
    Height (English) 96.5 in.
    Sash Opening (Metric) 25.4cm,25.4cm
    Amperage 12A
    Depth (Metric) 81.2cm
    Hertz 50/60Hz
    Voltage 100V
    Weight (English) 721 lb.
    Type Purifier Axiom C1 Biological Safety Cabinet

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit