Learn More
Labconco™ Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet
Labconco Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet features ergonomic Inclination™ Technology and MyLogic™ Operating System.
Manufacturer:  Labconco™ 304411020
 View more versions of this product
Catalog No. 30-441-1020
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
32 in.
100V, 50/60Hz, 12A, International
163cm
Fluorescent, UV
Safety Cabinet
636 lb.
54.2 in.
NSF International-Listed, ETL, Modified ASHRAE 110
25.4cm
81.2cm
Applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
Service fixture, Vacu-Pass Portal
N/A
10 in.
288.5kg
137.6cm
69 in.