Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet
Labconco Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet features ergonomic Inclination™ Technology and MyLogic™ Operating System.
Manufacturer: Labconco™ 304610300
Description
The Purifier Axiom Class II, Type C1 is first biological safety cabinet that can be used in recirculating Type A-mode for standard microbiological work or can be connected to exhaust system to function in Type-B mode for handling hazardous chemical vapors or radionuclides.
- Safe and easy to operate than traditional biosafety cabinets due to efficient, well-defined airflow pattern over its work surface
- Chem-Zone™(patent pending) increases chemical safety by creating large, clearly defined area within the enclosure
- Provides protection for personnel, product and laboratory environment by filtering hazardous particulates and aerosols, such as agents that require Biosafety Level 1, 2 or 3 containment, w/option of venting away hazardous fumes
Specifications
NSF International-Listed, ETL, Modified ASHRAE 110 | |
83.1cm | |
Applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards | |
245.1cm | |
Fluorescent | |
325w | |
Two GFCI duplex | |
25.4cm | |
684 CFM @ 0.2 in. s.p. | |
1000 lb. | |
78.2 in. |
32.7 in. | |
115V, 60Hz, 12A, Domestic | |
96.5 in. | |
Base stand and Bag-In/Bag-Out Exhaust Filter | |
North America | |
Safety Cabinet | |
10 in. | |
Class II Safety Cabinet | |
5 years | |
453.59kg | |
199cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title