Labconco™ Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet

Labconco Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet features ergonomic Inclination™ Technology and MyLogic™ Operating System.

Manufacturer: Labconco™ 304610150

 View more versions of this product

Catalog No. 30-461-0150

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Electrical Requirements 230V, 50/60Hz, 6A, International
    For Use With (Application) Applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
    Product Type Safety Cabinet
    Height (Metric) 245.1cm
    Plug Type British (UK)
    Certifications/Compliance NSF International-Listed, ETL, Modified ASHRAE 110,CE
    Width (English) 78.2 in.
    Width (Metric) 198.6cm
    Amperage 6A
    Depth (English) 32.6 in.
    Hertz 50/60Hz
    Includes Base stand
    Lighting Fluorescent
    Voltage 230V
    Weight (Metric) 453.6kg
    Height (English) 96.5 in.
    Depth (Metric) 82.8cm
    Sash Opening (English) 10 in.
    Sash Opening (Metric) 25.4cm,25.4cm
    Weight (English) 1000 lb.
    Type Purifier Axiom C1 Biological Safety Cabinet

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit