Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Purifier™ Cell Logic+™ Class II A2 Biosafety Cabinets, 3ft. Width, 9in. Opening, International Model

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

Manufacturer:  Labconco™ 322391030

 View more versions of this product

Catalog No. 32-239-1030


  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Add to cart

Specifications Description & Specifications

Specifications

6A
79.2cm
230V, 50/60Hz
27 in.
50/60Hz
Temp-Zone package, UV light, service fixtures, pass through
230V
585 lb.
42.3 in.
ADA, CE, NSF 49
61.8 in.
22.8cm
31.2 in.
LCD
Minute quantities of volatile toxic chemicals and tracer amounts of radionuclides as an adjunct to microbiological research
156.9cm
Powder Coated Steel
Fluorescent, UV
228 to 251 cfm
265.3kg
107.4cm
Stainless Steel
<63dBA
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit