Labconco™ Purifier™ Cell Logic+™ Class II A2 Biosafety Cabinets, 3ft. Width, 9in. Opening, International Model

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

Manufacturer: Labconco™ 332391040

 View more versions of this product

Catalog No. 33-239-1040

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ADA, CE, NSF 49,ADA, CE, NSF 49
    Depth (English) 31.2 in.
    Depth (Metric) 79.2cm
    Display LCD
    For Use With (Application) Minute quantities of volatile toxic chemicals and tracer amounts of radionuclides as an adjunct to microbiological research
    Height (English) Viewing 27 in.
    Height (Metric) 156.9cm
    Hertz 50/60Hz
    Housing Material Powder Coated Steel
    Includes Scope-Ready package, service fixtures, pass through
    Voltage 230V
    Volume 228 to 251 cfm
    Weight (English) 585 lb.
    Width (Metric) 107.4cm
    Height (English) 61.8 in.
    Noise Level <63dBA,<63dBA
    Sash Opening (Metric) 22.8cm,22.8cm
    Amperage 6A
    Chamber Material Stainless Steel,Stainless Steel
    Lighting Fluorescent
    Weight (Metric) 265.3kg
    Electrical Requirements 230V, 50/60Hz
    Width (English) 42.3 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit