Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Purifier™ Cell Logic+™ Class II A2 Biosafety Cabinets, 4ft. Width, 9in. Opening, International Model

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

Manufacturer:  Labconco™ 342491120

 View more versions of this product

Catalog No. 34-249-1120


  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Add to cart

Specifications Description & Specifications

Specifications

12A
79.2cm
100V, 50/60Hz
226.8 to 242cm
Powder Coated Steel Exterior and Stainless Steel Interior
Fluorescent
110V
303.9kg
137.9cm
Stainless Steel
<63dBA
31.2 in.
LCD
27 in.
50/60Hz
Scope-Ready package, Temp-Zone package, service fixtures, pass through, base stand
100V Japan
670 lb.
54.3 in.
CE, ETL, ADA, NSF 49, Class 5 ISO 14644-1, 2
89.3 to 95.3 in.
22.6cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit