Labconco™ Purifier™ Cell Logic+™ Class II A2 Biosafety Cabinets, 4ft. Width, 9in. Opening, International Model

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

Manufacturer: Labconco™ 322491120

 View more versions of this product

Catalog No. 32-249-1120

  Check Availability
Add to Cart

    Specifications

    Amperage 12A
    Depth (Metric) 79.2cm
    Electrical Requirements 100V, 50/60Hz
    Height (English) Viewing 27 in.
    Hertz 50/60Hz
    Includes Temp-Zone package, UV light, service fixtures, pass through, base stand
    Lighting Fluorescent, UV
    Plug Type 100V Japan
    Weight (English) 670 lb.
    Weight (Metric) 303.9kg
    Width (Metric) 137.9cm
    Certifications/Compliance CE, ETL, ADA, NSF 49, Class 5 ISO 14644-1, 2,CE, ETL, ADA, NSF 49, Class 5 ISO 14644-1, 2
    Noise Level <63dBA,<63dBA
    Sash Opening (Metric) 22.6cm,22.6cm
    Chamber Material Stainless Steel,Stainless Steel
    Depth (English) 31.2 in.
    Height (Metric) 226.8 to 242cm
    Housing Material Powder Coated Steel Exterior and Stainless Steel Interior
    Voltage 100V
    Width (English) 54.3 in.
    Height (English) 89.3 to 95.3 in.
    Display LCD

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit