Labconco™ Purifier™ Cell Logic+™ Class II A2 Biosafety Cabinets, 4ft. Width, 9in. Opening, International Model

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

Manufacturer: Labconco™ 322491070

 View more versions of this product

Catalog No. 32-249-1070

  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product.

Please contact customer service for assistance: 1-800-766-7000.

Add to Cart

    Specifications

    Height (English) Viewing 27 in.
    Hertz 50/60Hz
    Includes Temp-Zone package, UV light, service fixtures, pass through
    Lighting Fluorescent, UV
    Weight (English) 670 lb.
    Width (Metric) 137.9cm
    Chamber Material Stainless Steel,Stainless Steel
    Noise Level <63dBA,<63dBA
    Sash Opening (Metric) 22.6cm,22.6cm
    Amperage 6A
    Depth (Metric) 79.2cm
    Height (Metric) 156.9cm
    Housing Material Powder Coated Steel Exterior and Stainless Steel Interior
    Plug Type 230V India
    Width (English) 54.3 in.
    Height (English) 61.8 in.
    Display LCD
    Voltage 230V
    Certifications/Compliance CE, ETL, ADA, NSF 49, Class 5 ISO 14644-1, 2,CE, ETL, ADA, NSF 49, Class 5 ISO 14644-1, 2
    Depth (English) 31.2 in.
    Electrical Requirements 230V, 50/60Hz
    Weight (Metric) 303.9kg

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit