Labconco™ Purifier™ Cell Logic+™ Class II A2 Biosafety Cabinets, 5ft. Width, 9in. Opening

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

Manufacturer: Labconco™ 322591100

 View more versions of this product

Catalog No. 32-259-1100

  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product.

Please contact customer service for assistance: 1-800-766-7000.

Add to Cart

    Specifications

    Amperage 16A
    Depth (Metric) 79.2cm
    Height (English) Viewing 27 in.
    Housing Material Powder Coated Steel
    Lighting Fluorescent, UV
    Width (English) 66.3 in.
    Width (Metric) 168.4cm
    Chamber Material Stainless Steel,Stainless Steel
    Noise Level <63dBA,<63dBA
    Certifications/Compliance ADA, CAN/CSA C22.2, ETL, NSF 49, UL,ADA, CAN/CSA C22.2, ETL, NSF 49, UL
    Display LCD
    Height (Metric) 226.8 to 242cm
    Plug Type 115V Domestic
    Voltage 115V
    Weight (English) 715 lb.
    Height (English) 89.3 to 95.3 in.
    Depth (English) 31.2 in.
    Includes Temp-Zone package, UV light, service fixtures, pass through, base stand
    Sash Opening (Metric) 22.8cm,22.8cm
    Weight (Metric) 324.3kg

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit