Labconco™ Purifier™ Cell Logic+™ Class II A2 Biosafety Cabinets, 5ft. Width, 9in. Opening, International Model

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

Manufacturer: Labconco™ 342591150

 View more versions of this product

Catalog No. 34-259-1150

  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product.

Please contact customer service for assistance: 1-800-766-7000.

Add to Cart

    Specifications

    Depth (English) 31.2 in.
    Depth (Metric) 79.2cm
    Electrical Requirements 230V 50/60Hz
    For Use With (Application) Minute quantities of volatile toxic chemicals and tracer amounts of radionuclides as an adjunct to microbiological research
    Height (Metric) 226.8 to 242cm
    Hertz 50/60Hz
    Housing Material Powder Coated Steel
    Includes Scope-Ready package, Temp-Zone package, service fixtures, pass through, base stand
    Plug Type 230V UK
    Weight (English) 715 lb.
    Weight (Metric) 324.3kg
    Width (Metric) 168.4cm
    Certifications/Compliance ADA, CE, NSF 49,ADA, CE, NSF 49
    Chamber Material Stainless Steel,Stainless Steel
    Sash Opening (Metric) 22.8cm,22.8cm
    Amperage 8A
    Display LCD
    Height (English) Viewing 27 in.
    Lighting Fluorescent
    Noise Level <63dBA,<63dBA
    Voltage 230V
    Width (English) 66.3 in.
    Height (English) 89.3 to 95.3 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit