Labconco™ Purifier™ Cell Logic+™ Class II A2 Biosafety Cabinets, 6ft. Width, 9in. Opening, International Model

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

Manufacturer: Labconco™ 322691130

 View all options for this product

Catalog No. 32-269-1130

  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product.

Please contact customer service for assistance: 1-800-766-7000.

Add to Cart


    Amperage 8A
    Depth (English) 31.2 in.
    Depth (Metric) 79.2cm
    Certifications/Compliance CE, ETL, ADA, NSF 49, Class 5 ISO 14644-1, 2
    Chamber Material Stainless Steel
    Height (English) 89.3 to 95.3 in.
    Noise Level <63dBA
    Sash Opening (Metric) 22.6cm
    Display LCD
    Height (English) Viewing 27 in.
    Height (Metric) 226.8 to 242cm
    Housing Material Powder Coated Steel Exterior and Stainless Steel Interior
    Includes Temp-Zone package, UV light, service fixtures, pass through, base stand
    Lighting Fluorescent, UV
    Plug Type Schuko
    Voltage 230V
    Weight (English) 805 lb.
    Weight (Metric) 365.1kg
    Width (English) 78.3 in.
    Width (Metric) 198.8cm

    MyLogic™ operating system provides intuitive programming, color-coded menus, status messages and alarms with diagnostic troubleshooting — all using full sentences.

    • Same ECM blower technology as Purifier Logic™; uses Constant Airflow Profile™ (CAP) to maintain precise volume of air as filters load
    • As an additional safety feature, cabinet will continuously display HEPA filter life remaining and cabinet status
    • Provides personnel, product and environmental protection; suitable for work in Biosafety Levels 1, 2 and 3
    • Discharges HEPA-filtered exhaust air directly into lab, or into an exhaust system through optional Canopy Connection
    • Equipped to meet the specific needs of the cell culture industry
    • Available with optional Scope-Ready™ package, Temp-Zone™ package or both
    • Scope-Ready package allows the users microscope to be integrated into the biosafety cabinet
    • Temp-Zone package includes a temperature controlled area on the work surface
    Inclination Technology™ Suite includes:
    • 10° angled sash
    • Patented curved air grille with built-in armrest
    • Fully accessible controls and eye-level mounted color display with MyLogic operating system
    • Flush-mounted, slowly self-closing electrical outlet covers
    • ECM is quieter and cooler than comparable PSC motors
    • Large interior work area

    Some cabinets include non-welded base stands. For cabinets that do not include the base stand, one is required and is sold separately.

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit