Labconco™ Purifier™ Cell Logic+™ Class II A2 Biosafety Cabinets, 6ft. Width, 9in. Opening, International Model

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

Manufacturer: Labconco™ 332691070

 View more versions of this product

Catalog No. 33-269-1070

  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product.

Please contact customer service for assistance: 1-800-766-7000.

Add to Cart

    Specifications

    Amperage 8A
    Depth (English) 31.2 in.
    Height (English) Viewing 27 in.
    Housing Material Powder Coated Steel Exterior and Stainless Steel Interior
    Chamber Material Stainless Steel,Stainless Steel
    Height (English) 61.8 in.
    Noise Level <63dBA,<63dBA
    Depth (Metric) 79.2cm
    Display LCD
    Height (Metric) 156.9cm
    Lighting Fluorescent
    Plug Type India
    Voltage 230V
    Weight (Metric) 365.1kg
    Width (Metric) 198.8cm
    Certifications/Compliance CE, ETL, ADA, NSF 49, Class 5 ISO 14644-1, 2,CE, ETL, ADA, NSF 49, Class 5 ISO 14644-1, 2
    Includes Scope-ready package, service fixtures and pass through
    Sash Opening (Metric) 22.6cm,22.6cm
    Weight (English) 805 lb.
    Width (English) 78.3 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit