Labconco™ Purifier™ Cell Logic+™ Class II A2 Biosafety Cabinets, 6ft. Width, 9in. Opening, International Model

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

Manufacturer: Labconco™ 342691120

 View more versions of this product

Catalog No. 34-269-1120

  Check Availability
Add to Cart

    Specifications

    Display LCD
    Lighting Fluorescent
    Voltage 100V
    Weight (English) 805 lb.
    Amperage 12A
    Certifications/Compliance CE, ETL, ADA, NSF 49, Class 5 ISO 14644-1, 2,CE, ETL, ADA, NSF 49, Class 5 ISO 14644-1, 2
    Depth (English) 31.2 in.
    Depth (Metric) 79.2cm
    Height (English) Viewing 27 in.
    Height (Metric) 226.8 to 242cm
    Includes Scope-Ready package, Temp-Zone package, service fixtures, pass through, base stand
    Plug Type Japan
    Weight (Metric) 365.1kg
    Width (English) 78.3 in.
    Chamber Material Stainless Steel,Stainless Steel
    Height (English) 89.3 to 95.3 in.
    Noise Level <63dBA,<63dBA
    Housing Material Powder Coated Steel Exterior and Stainless Steel Interior
    Sash Opening (Metric) 22.6cm,22.6cm
    Width (Metric) 198.8cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit