Labconco™ Purifier™ Cell Logic+™ Class II B2 Biosafety Cabinets, 4ft. Width, 8in. Opening

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

Manufacturer: Labconco™ 343481010

 View more versions of this product

Catalog No. 34-348-1010

  Check Availability
Add to Cart

    Specifications

    Amperage 6A
    Electrical Requirements 230V 50/60Hz
    Height (English) Viewing 27 in.
    Housing Material Powder Coated Steel Exterior and Stainless Steel Interior
    Lighting Fluorescent, UV
    Plug Type 230V North America
    Width (Metric) 137.9cm
    Sash Opening (Metric) 20.3cm,20.3cm
    Depth (Metric) 79.2cm
    Display LCD
    Height (Metric) 184.4cm
    Hertz 50/60Hz
    Noise Level <63dBA,<63dBA
    Weight (English) 650 lb.
    Width (English) 54.3 in.
    Chamber Material Stainless Steel,Stainless Steel
    Height (English) 72.6 in.
    Certifications/Compliance ADA, CAN/CSA C22.2, CE, ETL, NSF 49, UL,ADA, CAN/CSA C22.2, CE, ETL, NSF 49, UL
    Depth (English) 31.2 in.
    Includes Scope-Ready package, Temp-Zone package, service fixtures, pass through
    Voltage 230V
    Weight (Metric) 294.8kg

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit