Labconco™ Purifier™ Cell Logic+™ Class II B2 Biosafety Cabinets, 4ft. Width, 8in. Opening, International Models

Biohazard cabinets with advanced ergonomic design provide user comfort and functionality

Manufacturer: Labconco™ 333481140

 View more versions of this product

Catalog No. 33-348-1140

  Check Availability
Add to Cart

    Specifications

    Depth (English) 31.2 in.
    Depth (Metric) 79.2cm
    Display LCD
    Electrical Requirements 230V 50/60Hz
    Hertz 50/60Hz
    Housing Material Powder Coated Steel
    Includes Scope-Ready package, service fixtures, pass through, base stand
    Plug Type China/Australia
    Voltage 230V
    Weight (Metric) 331.1kg
    Width (English) 54.3 in.
    Height (English) 100.1 to 106.1 in.
    Noise Level <63dBA,<63dBA
    Sash Opening (Metric) 20.3cm,20.3cm
    Amperage 6A
    Certifications/Compliance ADA, CE, NSF 49,ADA, CE, NSF 49
    Chamber Material Stainless Steel,Stainless Steel
    Height (English) Viewing 27 in.
    Height (Metric) 254.2 to 269.2cm
    Lighting Fluorescent
    Weight (English) 730 lb.
    Width (Metric) 137.9cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit