Labconco™ Purifier™ Cell Logic+™ Class II B2 Biosafety Cabinets, 4ft. Width, 8in. Opening, International Models

Biohazard cabinets with advanced ergonomic design provide user comfort and functionality

Manufacturer: Labconco™ 343481140

 View more versions of this product

Catalog No. 34-348-1140

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Depth (English) 31.2 in.
    Housing Material Powder Coated Steel
    Includes Scope-Ready package, Temp-Zone package, service fixtures, pass through, base stand
    Lighting Fluorescent, UV
    Weight (Metric) 331.1kg
    Width (Metric) 137.9cm
    Chamber Material Stainless Steel,Stainless Steel
    Amperage 6A
    Certifications/Compliance ADA, CE, NSF 49,ADA, CE, NSF 49
    Depth (Metric) 79.2cm
    Display LCD
    Height (Metric) 254.2 to 269.2cm
    Plug Type China/Australia
    Voltage 230V
    Weight (English) 730 lb.
    Width (English) 54.3 in.
    Height (English) 100.1 to 106.1 in.
    Noise Level <63dBA,<63dBA
    Height (English) Viewing 27 in.
    Sash Opening (Metric) 20.3cm,20.3cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit