Labconco™ Purifier™ Cell Logic+™ Class II B2 Biosafety Cabinets, 4ft. Width, 8in. Opening, International Models

Biohazard cabinets with advanced ergonomic design provide user comfort and functionality

Manufacturer: Labconco™ 323481130

 View more versions of this product

Catalog No. 32-348-1130

  Check Availability
Add to Cart

    Specifications

    Height (English) Viewing 27 in.
    Width (English) 54.3 in.
    Width (Metric) 137.9cm
    Housing Material Powder Coated Steel
    Weight (Metric) 331.1kg
    Amperage 6A
    Chamber Material Stainless Steel,Stainless Steel
    Depth (English) 31.2 in.
    Display LCD
    Height (Metric) 254.2 to 269.2cm
    Includes Temp-Zone package, UV light, service fixtures, pass through and base stand
    Plug Type Schuko
    Voltage 230V
    Weight (English) 730 lb.
    Certifications/Compliance ADA, CE, NSF 49,ADA, CE, NSF 49
    Height (English) 100.1 to 106.1 in.
    Depth (Metric) 79.2cm
    Lighting Fluorescent, UV
    Noise Level <63dBA,<63dBA
    Sash Opening (Metric) 20.3cm,20.3cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit