Labconco™ Purifier™ Cell Logic+™ Class II B2 Biosafety Cabinets, 4ft. Width, 8in. Opening, International Models

Biohazard cabinets with advanced ergonomic design provide user comfort and functionality

Manufacturer: Labconco™ 323481150

 View more versions of this product

Catalog No. 32-348-1150

  Check Availability
Add to Cart

    Specifications

    Amperage 6A
    Certifications/Compliance ADA, CE, NSF 49,ADA, CE, NSF 49
    Depth (English) 31.2 in.
    Depth (Metric) 79.2cm
    Display LCD
    Height (English) Viewing 27 in.
    Housing Material Powder Coated Steel
    Includes Temp-Zone package, UV light, service fixtures, pass through, base stand
    Lighting Fluorescent, UV
    Plug Type UK
    Voltage 230V
    Weight (English) 730 lb.
    Weight (Metric) 331.1kg
    Width (English) 54.3 in.
    Width (Metric) 137.9cm
    Chamber Material Stainless Steel,Stainless Steel
    Height (English) 100.1 to 106.1 in.
    Noise Level <63dBA,<63dBA
    Height (Metric) 254.2 to 269.2cm
    Sash Opening (Metric) 20.3cm,20.3cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit