Labconco™ Purifier™ Cell Logic+™ Class II B2 Biosafety Cabinets, 6ft. Width, 8in. Opening, International Models

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

Manufacturer: Labconco™ 333681150

 View more versions of this product

Catalog No. 33-368-1150

  Check Availability
Add to Cart

    Specifications

    Display LCD
    Lighting Fluorescent
    Voltage 230V
    Weight (Metric) 426.4kg
    Certifications/Compliance ADA, CE, NSF 49,ADA, CE, NSF 49
    Amperage 8A
    Chamber Material Stainless Steel,Stainless Steel
    Depth (English) 31.2 in.
    Depth (Metric) 79.2cm
    Height (English) Viewing 27 in.
    Height (Metric) 254.2 to 269.2cm
    Housing Material Powder Coated Steel
    Includes Scope-Ready package, service fixtures, pass through, base stand
    Plug Type UK
    Weight (English) 940 lb.
    Width (English) 78.3 in.
    Height (English) 100.1 to 106.1 in.
    Sash Opening (Metric) 20.3cm,20.3cm
    Width (Metric) 198.8cm
    Noise Level <63dBA,<63dBA

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit