Labconco™ Purifier™ Cell Logic+™ Class II B2 Biosafety Cabinets, 6ft. Width, 8in. Opening, International Models

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

Manufacturer: Labconco™ 343681030

 View more versions of this product

Catalog No. 34-368-1030

  Check Availability
Add to Cart

    Specifications

    Amperage 8A
    Chamber Material Stainless Steel,Stainless Steel
    Depth (English) 31.2 in.
    Depth (Metric) 79.2cm
    Display LCD
    Height (Metric) 184.4cm
    Housing Material Powder Coated Steel
    Includes Scope-Ready package, Temp-Zone package, service fixtures, pass through
    Lighting Fluorescent, UV
    Noise Level <63dBA,<63dBA
    Plug Type Schuko
    Weight (English) 840 lb.
    Width (Metric) 198.8cm
    Certifications/Compliance ADA, CE, NSF 49,ADA, CE, NSF 49
    Sash Opening (Metric) 20.3cm,20.3cm
    Height (English) Viewing 27 in.
    Voltage 230V
    Weight (Metric) 381kg
    Width (English) 78.3 in.
    Height (English) 72.6 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit