Learn More
Labconco™ Filters
Carbon-based and HEPA filters for use in Labconco enclosures which require periodic replacement
Manufacturer: Labconco™ 3707900
Description
Carbon-based and HEPA filters for use in Labconco enclosures which require periodic replacement
Replacement HEPA Filter, 99.99% efficient on particles 0.3 micron in size. Not suitable for use with biohazardous materials or volatile organic material. For use with Labconco FilterMate Portable Exhausters, Purifier Class I Enclosures, Purifier Filtered PCR Enclosures, Purifier HEPA Filtered Enclosures, Purifier Vertical Clean Benches, XPert Filtered Balance Stations, XPert and RXPert Filtered Balance Systems.Specifications
3.1 in. | |
10 to 20 x 18 x 18 in. (25.4 to 50.8 x 45.7 x 45.7cm) | |
HEPA | |
2, 3, 4 ft. Purifier™ class I safety, HEPA filtered, vertical clean bench and PCR enclosures | |
45.7cm | |
25.4 to 50.8cm | |
40 lb. | |
18 in. |
7.8cm | |
99.99% | |
Particulate | |
18 in. | |
10 to 20 in. | |
Filter | |
18.1kg | |
45.7cm |