Learn More
Labconco™ Filters
Carbon-based and HEPA filters for use in Labconco enclosures which require periodic replacement
$710.00 - $910.00
Specifications
Efficiency 99.99%
Filter Type HEPA
Product Type Filter
View More Specs
Products 2
Catalog Number Mfr. No. Length (English) Weight (English) Length (Metric) Weight (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Weight (English) Length (Metric) Weight (Metric) Price Quantity & Availability  
16-304-67
View Documents
Labconco™
3707900
10 to 20 in. 40 lb. 25.4 to 50.8cm 18.1kg
Each for $710.00
Add to cart
16-304-68
View Documents
Labconco™
3707901
30 to 40 in. 81 lb. 76.2 to 101.6cm 36.7kg
Each for $910.00
Add to cart
Description
Description
Carbon-based and HEPA filters for use in Labconco enclosures which require periodic replacement
Replacement HEPA Filter, 99.99% efficient on particles 0.3 micron in size. Not suitable for use with biohazardous materials or volatile organic material. For use with Labconco FilterMate Portable Exhausters, Purifier Class I Enclosures, Purifier Filtered PCR Enclosures, Purifier HEPA Filtered Enclosures, Purifier Vertical Clean Benches, XPert Filtered Balance Stations, XPert and RXPert Filtered Balance Systems.
Specifications
Specifications
99.99%
HEPA
Filter