Learn More
Labconco™ Purifier™ Fume Hood Prefilter and Blower for Purifier™ Trace-Metals Workstation
Supplier: Labconco™ 3727500
Description
For use with Labconco PVC Total Exhaust Clean Benches, Purifier Trace-Metals Workstation, Purifier Forensic Enclosure and Purifier Class 100 Chemical Station.
- Fiberglass Remote Exhaust Blower removes contaminated air from workstations or enclosures
- Prefilter is a replacement polyurethane foam filter traps large particles to extend life of HEPA filter (enclosure requires two)
Order Info
Ductwork not supplied.
Specifications
Prefilter | |
3 ft. Purifier™ trace-metals workstation, PVC total-exhaust clean benches, forensic enclosure and class 100 chemical station | |
Prefilter | |
Polyurethane Foam |