Labconco™ Purifier™ HEPA-Filtered Safety Enclosures with Guardian Monitor

Labconco Purifier™ HEPA-Filtered Safety Enclosures with Guardian Monitor provide personnel and environmental biohazard protection.

Manufacturer: Labconco™ 3980402

 View more versions of this product

Catalog No. 10-304-131

  Check Availability
Add to Cart

    Specifications

    Height (English) 37.08 in.
    Type Laminar flow safety cabinet
    Hertz 60Hz
    Volume 220 to 310 cfm
    Weight (English) 195 lb.
    Amperage 3A
    Electrical Requirements 115V 60Hz
    Filter Type HEPA
    Housing Material Epoxy Coated Aluminum and Steel
    Voltage 115V
    Weight (Metric) 88.45kg
    Width (English) 47.75 in.
    Sash Opening (Metric) 23.97cm,23.97cm
    Width (Metric) 121.28cm
    Lighting Fluorescent

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit