Labconco™ Purifier™ HEPA-Filtered Safety Enclosure, Without Guardian Monitor

Ensure personnel and environmental biohazard protection by outfitting your lab with the Labconco Purifier™ HEPA-Filtered Safety Enclosure.

Manufacturer: Labconco™ 3980400

 View all options for this product

Catalog No. 10-304-130

  Check Availability
Add to Cart


    Amperage 3A
    Height (English) 37.08 in.
    Sash Opening (Metric) 23.97cm
    Type Laminar Flow Cabinet
    Electrical Requirements 115V 60Hz
    Filter Type HEPA
    Hertz 60Hz
    Housing Material Epoxy Coated Aluminum and Steel
    Lighting Fluorescent
    Voltage 115V
    Volume 220 to 310 cfm
    Weight (English) 195 lb.
    Weight (Metric) 88.45kg
    Width (English) 47.75 in.
    Width (Metric) 121.28cm

    Meet requirements for Biosafety Level 1, 2, or 3 containment without product protection, making these an economical alternative to Class II biosafety cabinets. Also suitable for chemical and particulate powder weighing and other procedures generating fine dusts or aerosols. HEPA-filtered enclosures require no outside exhausting.

    • Total exhaust from 155 to 310cfm, depending on hood width
    • Sash pivots 10°, from 9.5 in. (24cm) operating position to 20 in. (50.8cm) opening for loading and maintenance
    • Quiet variable-speed, solid-state internal blower maintains appropriate air velocity
    • Airfoil with Clean-Sweep™ airflow openings
    • Front-mounted pressure gauge monitors pressure differential against the HEPA filter
    • HEPA filter is 99.99% efficient on 0.3µm particles (where applicable)
    • Negative pressure design ensures containment—should a leak occur, the system traps the contaminated air and recirculates it through the filter
    • Class I Enclosures have a 15w, 254nm germicidal UV lamp to use in conjunction with surface disinfection to ensure decontamination of biohazardous particulates while unattended

    • 0.25 in. (6.4mm) Tempered glass side panels and sash
    • Glare-free fluorescent lamp mounted outside contamination area
    • Blower and light switches are front-mounted for easy access
    • Powder-coated aluminum frame and steel rear plenum and zone-perforated rear baffle
    • Back panel has two utility ports for service connections
    • Open bottom allows enclosure to be placed on existing cabinetry or accessory dished work surface

    For operator safety, UV lamp cannot be turned on when fluorescent lamp is in use.

  • Description & Specifications
  • Accessories

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit