Labconco™ Purifier™ HEPA-Filtered Safety Enclosure, Without Guardian Monitor

Ensure personnel and environmental biohazard protection by outfitting your lab with the Labconco Purifier™ HEPA-Filtered Safety Enclosure.

Manufacturer: Labconco™ 3980220

 View more versions of this product

Catalog No. 10-304-124

  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product.

Please contact customer service for assistance: 1-800-766-7000.

Add to Cart

    Specifications

    Amperage 2A
    Housing Material Epoxy Coated Aluminum and Steel
    Voltage 230V
    Weight (English) 125 lb.
    Width (English) 23.75 in.
    Width (Metric) 60.32cm
    Height (English) 37.08 in.
    Sash Opening (Metric) 23.97cm,23.97cm
    Type Enclosure
    Hertz 50/60Hz
    Volume 110 to 155 cfm
    Weight (Metric) 57kg
    Electrical Requirements 230V 50/60Hz
    Filter Type HEPA
    Lighting Fluorescent

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit