Labconco™ Purifier™ Horizontal Clean Bench, 4ft.

When using a Labconco Purifier™ Horizontal Clean Bench, workers are protected from particulate contamination.

Manufacturer: Labconco™ 3888420

 View more versions of this product

Catalog No. 13-118-120

  N/A
Add to Cart

Alert

Units do not protect user and should not be used in conjunction with biohazardous material, toxins or radionuclides.


Required Accessories : Supporting base (sold separately).
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Depth (English) Interior 21 in.
    Width (English) Exterior 51 in.
    Height (Metric) Exterior 122.9cm
    Color Glacier White
    Depth (English) 34 in.
    Depth (Metric) 86.4cm
    Voltage 115V
    Plug Type Domestic
    Net Weight (English) 455 lb.
    Net Weight (Metric) 206kg
    Lighting Fluorescent, UV
    Ports Utility
    Filter Efficiency 99.99%
    Filter Type HEPA
    Amperage 15A
    Certifications/Compliance CAN/CSA C22.2, ETL, UL, CE
    Material Type 304 Brushed Stainless Steel
    Depth (Metric) Interior 53.3cm
    Weight (English) Shipping 455 lb.
    Width (Metric) Exterior 129.5cm
    Height (English) Exterior 48.4 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit