Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Purifier™ Horizontal Clean Bench, 5 ft. without UV Light

When using a Labconco Purifier™ Horizontal Clean Bench, workers are protected from particulate contamination.

Manufacturer:  Labconco™ 3250003

 View more versions of this product

Catalog No. 32-500-03


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

39 in.
230V 50/60Hz
26 in.
62 in.
48.4 in.
230V
260kg
99.99%
4A
Stainless Steel
99.1cm
Plant tissue culture, media plate preparation, electronics inspection, medical device assembly and pharmacy drug preparation
66cm
157.4cm
122.9cm
575 lb.
Fluorescent
HEPA
CAN/CSA C22.2, ETL, UL, CE
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit