Labconco™ Purifier™ Logic+™ Class II A2 Biosafety Cabinets, 5 ft. Width, 8 in. Opening, International Model

Protect from hazardous particulates such as agents requiring Biosafety Level 1, 2 or 3 containment

Manufacturer: Labconco™ 302581070

 View more versions of this product

Catalog No. 30-258-1070

  Check Availability
Add to Cart

    Specifications

    Depth (English) 31.2 in.
    Display LCD
    Height (Metric) 156.7cm
    Lighting Fluorescent, UV
    Voltage 230V
    Weight (English) 715 lb.
    Weight (Metric) 324.3kg
    Width (English) 66.3 in.
    Width (Metric) 168.4cm
    Certifications/Compliance ADA, CE, NSF 49, ETL,ADA, CE, NSF 49, ETL
    Height (English) 61.7 in.
    Noise Level <63dBA,<63dBA
    Sash Opening (Metric) 20.3cm,20.3cm
    Amperage 8A
    Chamber Material Stainless Steel,Stainless Steel
    Depth (Metric) 79.2cm
    Height (English) Viewing 27 in.
    Housing Material Powder Coated Steel Exterior and Stainless Steel Interior
    Includes UV light, service fixtures and pass through

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit