Learn More
Labconco™ Purifier™ Logic+ Class II B2 Biosafety Cabinets, 4 ft. Width, 8 in. Opening, International Model
Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality
Supplier: Labconco™ 303481120
Description
MyLogic™ operating system provides intuitive programming, color-coded menus, status messages and alarms with diagnostic troubleshooting — all using full sentences.
- ECM blower technology combined with CAP (Constant Airflow Profile™) maintains precise volume of air as filters load
- Interior-mounted LCD displays HEPA filter life remaining and cabinet status
- Provides personnel, product and environmental protection; suitable for work in Biosafety Levels 1, 2 and 3
- Cabinets are designed to be hard ducted to the outside
- Flush-mounted stainless steel electrical receptacle covers with dampening hinges
- Patented curved air grille with built-in armrest
- Fully accessible controls and eye-level mounted color display with MyLogic operating system
Some models have base stands, UV lights, service fixtures, pass throughs and/or a combination of those features included. (Refer to nominal width, sash opening, electrical configuration and product specifications for details.)
Specifications
ADA, NSF 49 | |
International | |
20.3 cm | |
Powder Coated Steel Exterior and Stainless Steel Interior | |
Fluorescent, UV | |
54.3 in. | |
27 in. | |
79.2 cm | |
331.1 Kg | |
12 A |
Stainless Steel | |
100.1 to 106.1 in. | |
LCD | |
Base Stand, UV Light, Service Fixtures, Pass-through | |
254.2 to 269.2 cm | |
137.9 cm | |
31.2 in. | |
730 lb. | |
100 V |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.