Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ RapidVap™ Vetex+ Dry Evaporator Blocks

Required for use of the RapidVap Dry Block Evaporator

Manufacturer:  Labconco™ 7321300

 View more versions of this product

Catalog No. 10-269-677


  Check Availability
Add to cart

Specifications

Description

  • Comprised of solid aluminum with stainless steel handles for lifting out of evaporator
  • Comes with 0.21 in. (0.5cm) diameter port in upper left hand corner for insertion of temperature probe to monitor upper block temperature
  • Multiple block variations available to suit tube size
  • Custom blocks also available
Description & Specifications

Specifications

RapidVap vertex dry evaporator
0.21 in.
Fifty 13 x 100mm tube
50.0
50
Aluminum
5mm
Dry evaporator block
8mL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit