Learn More
Labconco™ Replacement Disposable Prefilters
SDP
Prefilters for Vertical Clean Benches and Filtered PCR Enclosures
Supplier:  Labconco™ 3935702
 View more versions of this product
Catalog No. 16-108-282
Add to Cart
Description
Description
  • Corrugated paper prefilters for Purifier Vertical Clean Benches and Filtered PCR Enclosures trap large particles to extend HEPA filter life
Specifications
Specifications
Prefilter
1 in.
0.5 Kg
16.5 in.
102.9 cm
Prefilter
4 ft. Purifier™ filtered PCR enclosures and vertical clean benches manufactured before 8/2004 and newer
1 lb.
40.5 in.
2.5 cm
41.9 cm
Product Suggestions
Product Suggestions