Learn More
Labconco™ Replacement Door Filters for Protector Evidence Drying Cabinets
SDP
Precut to fit the dimensions of the door.
Supplier:  Labconco™ 3872602
Catalog No. 10-359-253
Add to Cart
Description
Description
Polyester prefilters trap larger particles in air entering the cabinet. MERV 11 rating. Precut to fit the door of Protector Evidence Drying Cabinets. Package of 6.
Specifications
Specifications
Prefilter
1 lb.
Polyester
5 ft. Protector™ Evidence Drying Cabinet
0.5 Kg