Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco Replacement Filters for Benchtop Fume Adsorbers
Replacement filters for use with Labconco Fume Adsorbers and Fume Adsorber-2s.
$569.00 - $586.00
Specifications
Weight (English) | 11 lb. |
---|---|
Filter Type | Carbon |
For Use With (Equipment) | Benchtop fume adsorber |
Product Type | Filter |
Weight (Metric) | 5kg |
Catalog Number | Mfr. No. | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Price | Quantity & Availability | |||||
10-090-104
|
Labconco
6961300 |
Organic vapor |
Each for $586.00
|
|
10-090-105
|
Labconco
6961301 |
Formaldehyde-formalin |
Each for $586.00
|
|
10-090-128
|
Labconco
6961302 |
Ammonia-amine |
Each for $569.00
|
|
Description
Replacement Ammonia Carbon Filter for use with ammonia and low molecular weight amines. 8.8 pounds of impregnated carbon. Adsorption weight 10% (0.88 lbs.). Adsorption volume 800 ml of 50% ammonia. For use with Labconco Fume Adsorber and Fume Adsorber-2. Fume Adsorber Models 3955202 and 3955222 include one filter; Fume Adsorber-2 Models 3955402 and 3955422 include two filters.Specifications
11 lb. | |
Benchtop fume adsorber | |
5kg |
Carbon | |
Filter |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title