Learn More
Labconco™ Replacement Filters for Benchtop Fume Adsorbers
SDP
Replacement filters for use with Labconco Fume Adsorbers and Fume Adsorber-2s.
Supplier:  Labconco™ 6961301
 View more versions of this product
Catalog No. 10-090-105
Add to Cart
Description
Description
Replacement Formaldehyde Carbon Filter for use with formaldehyde and gluteraldehyde. 7.5 pounds of impregnated carbon. Adsorption weight 10% (0.75 lb.). Adsorption volume 850 ml of 37% formalin. For use with Labconco Fume Adsorber and Fume Adsorber-2. Fume Adsorber Models 3955201 and 3955221 include one filter; Fume Adsorber-2 Models 3955401 and 3955421 include two filters.
Specifications
Specifications
Carbon
11 lb.
Filter
Benchtop Fume Adsorber
5 Kg
Formaldehyde-formalin
Product Suggestions
Product Suggestions