Learn More
Labconco™ Replacement Supply HEPA Filter for Class II Biosafety Cabinets
SDP
Replacement Supply HEPA Filter for Labconco Class II Biosafety Cabinets.
Supplier:  Labconco™ 3838401
 View more versions of this product
Catalog No. 10-269-295
Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative.
Description
Description
Replacement Supply HEPA Filter for Labconco Class II Biosafety Cabinets.
Replacement Supply HEPA Filter for Labconco Logic+ and Axiom Class II Biosafety Cabinets and PuriCare Procedure Stations.
Specifications
Specifications
0.9999
4 ft. Purifier™ Logic™ biosafety cabinet and Purifier™ Cell Logic™ biosafety cabinet
70 lb.
40 to 50 in.
18 in.
101.6 to 127 cm
Borosilicate Microfiber
48 in.
3.06 in.
HEPA
3.1 in.
31.8 Kg
48 in.
121.9 cm
45.7 cm
18 in.
Particulate