Learn More
Labconco™ Replacement Supply HEPA Filter for Class II Biosafety Cabinets
Replacement Supply HEPA Filter for Labconco Class II Biosafety Cabinets.
Manufacturer: Labconco™ 3838403
Description
Replacement Supply HEPA Filter for Labconco Class II Biosafety Cabinets.
Replacement Supply HEPA Filter for Labconco Logic+ and Axiom Class II Biosafety Cabinets and PuriCare Procedure Stations.Specifications
0.9999 | |
Particulate | |
3.06 in. | |
72 in. | |
105 lb. | |
6 ft. | |
183cm |
HEPA | |
6 ft. Purifier™ Logic™ biosafety cabinet and Purifier™ Cell Logic™ biosafety cabinet | |
72 in. | |
Borosilicate Microfiber | |
47.6kg | |
18 in. |