Learn More
Labconco™ ScrubAir™ Pipette Washer / Dryer Accessory, Spindle Kit
Easily adapt your ScrubAir glassware washer / dryer to permit cleaning of small test tubes or vials with the Labconco ScrubAir Spindle Kit.
Manufacturer:  Labconco™ 4615700
Catalog No. 10-359-222
Add to cart
Description
Description
  • Outfits small test tubes or vials, 7 mm ID or larger, to be washed in the ScrubAir
  • Kit contains 24 each 5mm OD polystyrene plastic spindles with caps and stainless steel test tube rack to hold up to 20 each test tubes
  • Shipping weight: 0.9 kg (2.0 lbs.)
Specifications
Specifications
ScrubAir Pipette Washer/Dryer
Kit