Labconco™ Shelves for FreeZone™ Freeze-Dry Systems

Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications

Manufacturer: Labconco™ 7441700

 View more versions of this product

Catalog No. 10-030-153

  Check Availability
Add to Cart

    Specifications

    Diameter (English) 10 in.
    Diameter (Metric) 25.4cm
    For Use With (Application) Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications
    For Use With (Equipment) Freezone™ drying chambers
    Height (English) 12.2 in.
    Height (Metric) 30.98cm
    Includes Three aluminum shelves with 0.25 in. rims, three stainless steel support rods, nine clips for shelf height adjustment, and three rubber feet
    Size Large
    Weight (English) 2 lb.
    Weight (Metric) 0.9kg

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit