Learn More
Labconco™ Single-Use Door Prefilters for 4 Ft. Evidence Drying Cabinets
May be retained for trace evidence. For use with Protector Evidence Drying Cabinets.
Manufacturer: Labconco™ 3951822
Description
Single-Use Door Evidence Prefilters may be examined and/or retained for trace evidence. Single-use. Polyester. Pre-cut to fit the door prefilter frame. Package of 10.Specifications
Prefilter | |
4 ft. Protector™ evidence drying cabinet | |
Polyester |