Labconco™ SpillStopper™ Work Surface for 6 Ft. Protector™Premier™and Protector™XL Hoods
Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.
Manufacturer: Labconco™ 9500600
 View more versions of this product
Catalog No. 10-369-161
  Check Availability
Add to Cart
    Specifications
    Material Epoxy resin
    Product Type No cutouts
  • Description & Specifications
Related Products
Product Content Feedback