Learn More
Labconco™ Stainless-Steel Work Surface for XPert™ Filtered Balance Stations and System
Choose an easy-to-maintain worksurface. The Labconco Stainless-Steel Work Surface is ideal for use with the XPert™ Filtered Balance Station and System.
Manufacturer:  Labconco™ 3970602
 View more versions of this product
Catalog No. 10-305-205
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
29 in.
73.6cm
2 ft. width Enclosure
2cm
Type 304 Stainless Steel
70 lb.
31.7kg
Without Cutout
2 ft. width Enclosure
0.8 in.
Front spill trough
Work Surface
24 in.
61cm